(4-(Tert-butyl)-2-chlorophenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

63879-5G 63879-1G 63879-25G 63879-500MG
77633-658EA 1441 USD
77633-658 77639-870 77656-758 77651-876
(4-(Tert-butyl)-2-chlorophenyl)boronic acid ≥95%
(4-(Tert-butyl)-2-chlorophenyl)boronic acid

Precaución: For Research Use Only

Formula: 212.4809 g/mol
MW: C₁₀H₁₄BClO₂
Storage Temperature: Ambient
MDL Number: MFCD34761557
CAS Number: 2756573-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR