2,2,2-Trifluoro-1-(4-hydroxy-2-methylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

63815-1G 63815-5G 63815-500MG
77635-262EA 1097 USD
77635-262 77656-756 77647-856
2,2,2-Trifluoro-1-(4-hydroxy-2-methylphenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(4-hydroxy-2-methylphenyl)ethanone

Precaución: For Research Use Only

Formula: 204.1458 g/mol
MW: C₉H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34169040
CAS Number: 1554935-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR