1-(4-(Benzyloxy)-3-(tert-butyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

63718-5G 63718-1G 63718-500MG
77630-560EA 1604 USD
77630-560 77643-888 77651-872
1-(4-(Benzyloxy)-3-(tert-butyl)phenyl)ethanone ≥95%
1-(4-(Benzyloxy)-3-(tert-butyl)phenyl)ethanone

Precaución: For Research Use Only

Formula: 282.3767 g/mol
MW: C₁₉H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD31555992
CAS Number: 146852-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR