1-(3-(Tert-butyl)-4-ethoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

63671-1G 63671-5G 63671-500MG
77639-868EA 638 USD
77639-868 77660-206 77651-870
1-(3-(Tert-butyl)-4-ethoxyphenyl)ethanone ≥95%
1-(3-(Tert-butyl)-4-ethoxyphenyl)ethanone

Precaución: For Research Use Only

Formula: 220.3074 g/mol
MW: C₁₄H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD22003879
CAS Number: 1498730-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR