1-(3-(Tert-butyl)-4-methoxyphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

63621-5G 63621-1G 63621-25G 63621-500MG
77630-558EA 1441 USD
77630-558 77639-866 77656-750 77647-854
1-(3-(Tert-butyl)-4-methoxyphenyl)-2,2,2-trifluoroethanone ≥95%
1-(3-(Tert-butyl)-4-methoxyphenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: C₁₃H₁₅F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD07380812
CAS Number: 1368534-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR