1-(2-Chloro-6-(methoxymethoxy)phenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

63614-5G 63614-25G 63614-1G 63614-500MG
77633-656EA 886 USD
77633-656 77656-748 77647-850 77647-852
1-(2-Chloro-6-(methoxymethoxy)phenyl)-2,2,2-trifluoroethanone ≥97%
1-(2-Chloro-6-(methoxymethoxy)phenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 268.6169 g/mol
MW: C₁₀H₈ClF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34761451
CAS Number: 2807466-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR