Ethyl 3-bromo-4-hydroxy-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

63532-5G 63532-1G 63532-500MG 63532-25G
77633-654EA 1398 USD
77633-654 77639-864 77653-056 77656-744
Ethyl 3-bromo-4-hydroxy-5-methylbenzoate ≥97%
Ethyl 3-bromo-4-hydroxy-5-methylbenzoate

Precaución: For Research Use Only

Formula: C₁₀H₁₁BrO₃
Storage Temperature: Ambient
MDL Number: MFCD34761406
CAS Number: 2092777-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR