1-(6-Bromo-3-chloro-2,4-difluorophenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

63471-5G 63471-25G 63471-1G 63471-500MG
77630-554EA 1441 USD
77630-554 77656-742 77643-886 77647-848
1-(6-Bromo-3-chloro-2,4-difluorophenyl)-2,2,2-trifluoroethanone ≥95%
1-(6-Bromo-3-chloro-2,4-difluorophenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 323.4419 g/mol
MW: C₈HBrClF₅O
Storage Temperature: Ambient
MDL Number: MFCD34761362
CAS Number: 2901066-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR