1-(2,6-Dichloro-3-methoxyphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

63463-5G 63463-25G 63463-1G 63463-500MG
77630-550EA 1441 USD
77630-550 77660-202 77643-882 77651-866
1-(2,6-Dichloro-3-methoxyphenyl)-2,2,2-trifluoroethanone ≥95%
1-(2,6-Dichloro-3-methoxyphenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 273.0360 g/mol
MW: C₉H₅Cl₂F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD28360083
CAS Number: 2090437-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR