1-(4-(Benzyloxy)phenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

63432-5G 63432-25G 63432-1G 63432-500MG
77635-258EA 264 USD
77635-258 77656-740 77643-880 77651-864
1-(4-(Benzyloxy)phenyl)ethanol ≥95%
1-(4-(Benzyloxy)phenyl)ethanol

Precaución: For Research Use Only

Formula: C₁₅H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD06654195
CAS Number: 36438-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR