1-(3-Ethyl-5-(methylthio)phenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

63343-5G 63343-1G 63343-25G 63343-500MG
77635-256EA 588 USD
77635-256 77639-860 77656-738 77651-862
1-(3-Ethyl-5-(methylthio)phenyl)-2,2,2-trifluoroethanone ≥97%
1-(3-Ethyl-5-(methylthio)phenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 248.2646 g/mol
MW: C₁₁H₁₁F₃OS
Storage Temperature: Ambient
MDL Number: MFCD34761301
CAS Number: 2764730-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR