1-(2-Chloro-4-methoxy-5-methylphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

62888-1G 62888-5G 62888-500MG
77639-850EA 531 USD
77639-850 77656-726 77651-854
1-(2-Chloro-4-methoxy-5-methylphenyl)-2,2,2-trifluoroethanone ≥97%
1-(2-Chloro-4-methoxy-5-methylphenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 252.6175 g/mol
MW: C₁₀H₈ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34761102
CAS Number: 2807448-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR