2-(Benzyloxy)-3-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

62503-5G 62503-1G 62503-25G 62503-500MG
77633-648EA 698 USD
77633-648 77639-848 77660-190 77651-850
2-(Benzyloxy)-3-iodopyridine ≥97%
2-(Benzyloxy)-3-iodopyridine

Precaución: For Research Use Only

Formula: 311.1183 g/mol
MW: C₁₂H₁₀INO
Storage Temperature: Ambient
MDL Number: MFCD30470512
CAS Number: 1816294-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR