5-Bromo-2-ethynyl-3-methylpyridine ≥95%

Supplier: AOBChem USA

Warning

623848-5G 623848-1G 623848-500MG
77656-716EA 2787 USD
77656-716 77643-870 77651-846
5-Bromo-2-ethynyl-3-methylpyridine ≥95%
5-Bromo-2-ethynyl-3-methylpyridine

Precaución: For Research Use Only

Formula: 196.0439 g/mol
MW: C₈H₆BrN
Storage Temperature: Ambient
CAS Number: 1211588-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR