3-Methyl-5-(2,2,2-trifluoroacetyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62320-5G 62320-1G 62320-25G 62320-500MG
77633-646EA 1266 USD
77633-646 77639-844 77656-714 77651-842
3-Methyl-5-(2,2,2-trifluoroacetyl)benzaldehyde ≥97%
3-Methyl-5-(2,2,2-trifluoroacetyl)benzaldehyde

Precaución: For Research Use Only

Formula: 216.1565 g/mol
MW: C₁₀H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34760798
CAS Number: 2764729-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR