1-(3-Bromo-5-chloro-2-fluorophenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

62270-5G 62270-1G 62270-25G 62270-500MG
77630-544EA 1441 USD
77630-544 77639-840 77656-712 77647-824
1-(3-Bromo-5-chloro-2-fluorophenyl)ethanol ≥95%
1-(3-Bromo-5-chloro-2-fluorophenyl)ethanol

Precaución: For Research Use Only

Formula: 253.4959 g/mol
MW: C₈H₇BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34814170
CAS Number: 2918967-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR