1-(3,6-Dimethoxypyridazin-4-yl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

62226-5G 62226-1G 62226-25G 62226-500MG
77633-644EA 998 USD
77633-644 77639-838 77656-710 77647-822
1-(3,6-Dimethoxypyridazin-4-yl)-2,2,2-trifluoroethanone ≥97%
1-(3,6-Dimethoxypyridazin-4-yl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: C₈H₇F₃N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34760767
CAS Number: 2355652-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR