2,2,2-Trifluoro-1-(4-methoxy-2,6-dimethylphenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
62097-5G
62097-1G
62097-25G
62097-500MG
77633-642EA
865
USD
77633-642
77639-836
77660-186
77651-840
2,2,2-Trifluoro-1-(4-methoxy-2,6-dimethylphenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(4-methoxy-2,6-dimethylphenyl)ethanone
Precaución: For Research Use Only
Formula:
C₁₁H₁₁F₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD22688434 CAS Number: 1417460-98-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...