3,4-Dibromo-6-chloro-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

61879-5G 61879-1G 61879-25G 61879-500MG
77633-640EA 1092 USD
77633-640 77639-834 77656-704 77651-838
3,4-Dibromo-6-chloro-1H-indazole ≥95%
3,4-Dibromo-6-chloro-1H-indazole

Precaución: For Research Use Only

Formula: 310.3731 g/mol
MW: C₇H₃Br₂ClN₂
Storage Temperature: Ambient
MDL Number: MFCD07781874
CAS Number: 887568-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR