2-Bromo-9-ethyl-9H-carbazole ≥97%

Supplier: AOBChem USA

Warning

617122-5G 617122-1G 617122-25G 617122-500MG
77630-542EA 1441 USD
77630-542 77639-832 77656-702 77651-834
2-Bromo-9-ethyl-9H-carbazole ≥97%
2-Bromo-9-ethyl-9H-carbazole

Precaución: For Research Use Only

Formula: C₁₄H₁₂BrN
Storage Temperature: Ambient
CAS Number: 56166-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR