4'-Bromo-3-nitro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

616639-5G 616639-25G 616639-1G 616639-500MG
77630-540EA 1441 USD
77630-540 77636-184 77643-866 77651-832
4'-Bromo-3-nitro-1,1'-biphenyl ≥97%
4'-Bromo-3-nitro-1,1'-biphenyl

Precaución: For Research Use Only

Formula: 278.1014 g/mol
MW: C₁₂H₈BrNO₂
Storage Temperature: Ambient
CAS Number: 32858-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR