2-Bromo-4-fluoro-3-iodo-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

616617-5G 616617-1G 616617-500MG
77660-184EA 2245 USD
77660-184 77643-864 77651-830
2-Bromo-4-fluoro-3-iodo-1-methylbenzene ≥97%
2-Bromo-4-fluoro-3-iodo-1-methylbenzene

Precaución: For Research Use Only

Formula: 314.9214 g/mol
MW: C₇H₅BrFI
Storage Temperature: Ambient
CAS Number: 1804910-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR