2-(2,4-Dichlorophenoxy)aniline ≥97%

Supplier: AOBChem USA

Warning

614979-5G 614979-1G 614979-25G 614979-500MG
77633-638EA 613 USD
77633-638 77639-830 77656-698 77651-828
2-(2,4-Dichlorophenoxy)aniline ≥97%
2-(2,4-Dichlorophenoxy)aniline

Precaución: For Research Use Only

Formula: 254.1119 g/mol
MW: C₁₂H₉Cl₂NO
Storage Temperature: Ambient
CAS Number: 26306-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR