2,3,6-Trichloroisonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

60721-5G 60721-1G 60721-25G 60721-500MG
77635-244EA 594 USD
77635-244 77639-812 77656-682 77651-814
2,3,6-Trichloroisonicotinic acid ≥97%
2,3,6-Trichloroisonicotinic acid

Precaución: For Research Use Only

Formula: 226.4445 g/mol
MW: C₆H₂Cl₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD34759903
CAS Number: 2764733-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR