(4-Bromonaphthalen-1-yl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

60662-5G 60662-1G 60662-25G 60662-500MG
77633-636EA 761 USD
77633-636 77639-808 77660-176 77647-806
(4-Bromonaphthalen-1-yl)(methyl)sulfane ≥97%
(4-Bromonaphthalen-1-yl)(methyl)sulphane

Precaución: For Research Use Only

Formula: 253.158 g/mol
MW: C₁₁H₉BrS
Storage Temperature: Ambient
MDL Number: MFCD30496776
CAS Number: 63279-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR