5-Bromo-3-chlorothiophene-2-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

606538-5G 606538-1G 606538-25G 606538-500MG
77633-634EA 671 USD
77633-634 77639-806 77656-678 77651-812
5-Bromo-3-chlorothiophene-2-carbaldehyde ≥97%
5-Bromo-3-chlorothiophene-2-carbaldehyde

Precaución: For Research Use Only

Formula: C₅H₂BrClOS
Storage Temperature: Ambient
CAS Number: 1503406-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR