5-Bromo-3-chlorothiophene-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

606535-5G 606535-500MG 606535-1G 606535-25G
77633-632EA 1412 USD
77633-632 77653-050 77639-804 77656-676
5-Bromo-3-chlorothiophene-2-carboxylic acid ≥97%
5-Bromo-3-chlorothiophene-2-carboxylic acid

Precaución: For Research Use Only

Formula: C₅H₂BrClO₂S
MW: 241.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22380014
CAS Number: 842135-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR