3-(Ethoxymethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

606391-5G 606391-1G 606391-25G 606391-500MG
77633-630EA 1357 USD
77633-630 77639-802 77660-174 77651-810
3-(Ethoxymethyl)benzaldehyde ≥97%
3-(Ethoxymethyl)benzaldehyde

Precaución: For Research Use Only

Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
CAS Number: 874513-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR