3,4,6-Tribromo-2-methylaniline ≥97%

Supplier: AOBChem USA

Warning

60611-5G 60611-25G
77635-242EA 105 USD
77635-242 77660-172
3,4,6-Tribromo-2-methylaniline ≥97%
3,4,6-Tribromo-2-methylaniline

Precaución: For Research Use Only

Formula: 343.841 g/mol
MW: C₇H₆Br₃N
Storage Temperature: Ambient
MDL Number: MFCD34759850
CAS Number: 857007-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR