1-(4-Chloro-3-isopropoxyphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

60474-5G 60474-1G 60474-25G 60474-500MG
77633-628EA 1166 USD
77633-628 77639-796 77660-168 77651-806
1-(4-Chloro-3-isopropoxyphenyl)-2,2,2-trifluoroethanone ≥97%
1-(4-Chloro-3-isopropoxyphenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: C₁₁H₁₀ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34759765
CAS Number: 2764729-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR