1-(4-Chloro-2-isopropoxyphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

60375-1G 60375-5G 60375-500MG
77639-794EA 556 USD
77639-794 77656-672 77651-804
1-(4-Chloro-2-isopropoxyphenyl)-2,2,2-trifluoroethanone ≥97%
1-(4-Chloro-2-isopropoxyphenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 266.644 g/mol
MW: C₁₁H₁₀ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34759715
CAS Number: 2764731-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR