(5-(4-Bromophenoxy)-2-fluorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

60365-5G 60365-1G 60365-25G 60365-500MG
77630-520EA 1441 USD
77630-520 77639-792 77656-670 77651-802
(5-(4-Bromophenoxy)-2-fluorophenyl)methanol ≥95%
(5-(4-Bromophenoxy)-2-fluorophenyl)methanol

Precaución: For Research Use Only

Formula: C₁₃H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34759709
CAS Number: 2807471-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR