2-(Difluoromethoxy)-1-iodo-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

60139-5G 60139-25G 60139-1G 60139-500MG
77630-518EA 1441 USD
77630-518 77660-166 77643-852 77647-796
2-(Difluoromethoxy)-1-iodo-3-methoxybenzene ≥97%
2-(Difluoromethoxy)-1-iodo-3-methoxybenzene

Precaución: For Research Use Only

Formula: 300.041 g/mol
MW: C₈H₇F₂IO₂
Storage Temperature: Ambient
CAS Number: 3037705-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR