2-Isopropoxy-3-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

60130-5G 60130-25G 60130-1G 60130-500MG
77635-240EA 436 USD
77635-240 77660-164 77643-850 77647-794
2-Isopropoxy-3-methoxybenzoic acid ≥95%
2-Isopropoxy-3-methoxybenzoic acid

Precaución: For Research Use Only

Formula: C₁₁H₁₄O₄
Storage Temperature: Ambient
MDL Number: MFCD12028129
CAS Number: 85686-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR