3-(Benzyloxy)isonicotinaldehyde ≥95%

Supplier: AOBChem USA

Warning

60114-1G 60114-5G 60114-500MG
77635-238EA 895 USD
77635-238 77656-668 77647-792
3-(Benzyloxy)isonicotinaldehyde ≥95%
3-(Benzyloxy)isonicotinaldehyde

Precaución: For Research Use Only

Formula: C₁₃H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD18256125
CAS Number: 1211517-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR