N-(2-(2-Hydroxypropyl)phenyl)pivalamide ≥97%

Supplier: AOBChem USA

Warning

601079-5G 601079-25G 601079-1G 601079-500MG
77630-516EA 1441 USD
77630-516 77636-178 77643-848 77647-790
N-(2-(2-Hydroxypropyl)phenyl)pivalamide ≥97%
N-(2-(2-Hydroxypropyl)phenyl)pivalamide

Precaución: For Research Use Only

Formula: 235.3220 g/mol
MW: C₁₄H₂₁NO₂
Storage Temperature: Ambient
CAS Number: 68965-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR