Ethyl 4-fluoro-3-formylbenzoate ≥97%

Supplier: AOBChem USA

Warning

59991-5G 59991-500MG 59991-1G 59991-25G
77635-234EA 342 USD
77635-234 77653-048 77639-788 77656-666
Ethyl 4-fluoro-3-formylbenzoate ≥97%
Ethyl 4-fluoro-3-formylbenzoate

Precaución: For Research Use Only

Formula: 196.175 g/mol
MW: C₁₀H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD28795857
CAS Number: 1806331-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR