1-Bromo-2-butyl-3-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

598862-5G 598862-1G 598862-25G 598862-500MG
77630-510EA 1770.57 USD
77630-510 77639-782 77656-662 77651-798
1-Bromo-2-butyl-3-fluorobenzene ≥97%
1-Bromo-2-butyl-3-fluorobenzene

Precaución: For Research Use Only

Formula: 231.1046 g/mol
MW: C₁₀H₁₂BrF
Storage Temperature: Ambient
CAS Number: 1341833-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR