4-(4-Fluorophenyl)-1-butene ≥97%

Supplier: AOBChem USA

Warning

598855-5G 598855-1G 598855-25G 598855-500MG
77635-232EA 545 USD
77635-232 77639-780 77660-158 77647-784
4-(4-Fluorophenyl)-1-butene ≥97%
4-(4-Fluorophenyl)-1-butene

Precaución: For Research Use Only

Formula: C₁₀H₁₁F
Storage Temperature: Ambient
MDL Number: MFCD00671360
CAS Number: 2248-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR