Ethyl 3-(2-bromo-6-fluoro-4-methylphenyl)-3-hydroxypropanoate ≥97%

Supplier: AOBChem USA

Warning

59699-5G 59699-25G 59699-1G 59699-500MG
77633-626EA 642 USD
77633-626 77660-156 77643-846 77651-796
Ethyl 3-(2-bromo-6-fluoro-4-methylphenyl)-3-hydroxypropanoate ≥97%
Ethyl 3-(2-bromo-6-fluoro-4-methylphenyl)-3-hydroxypropanoate

Precaución: For Research Use Only

Formula: 305.14 g/mol
MW: C₁₂H₁₄BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD34759347
CAS Number: 2755722-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR