(3-Ethyl-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

59561-5G 59561-1G 59561-25G 59561-500MG
77633-624EA 1441 USD
77633-624 77639-778 77656-660 77647-782
(3-Ethyl-5-methylphenyl)(methyl)sulfane ≥97%
(3-Ethyl-5-methylphenyl)(methyl)sulphane

Precaución: For Research Use Only

Formula: 166.283 g/mol
MW: C₁₀H₁₄S
Storage Temperature: Ambient
MDL Number: MFCD22489963
CAS Number: 1824582-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR