2-Bromo-7-methoxybenzofuran ≥95%

Supplier: AOBChem USA

Warning

592326-5G 592326-1G 592326-500MG
77656-654EA 2348 USD
77656-654 77643-844 77647-778
2-Bromo-7-methoxybenzofuran ≥95%
2-Bromo-7-methoxybenzofuran

Precaución: For Research Use Only

Formula: C₉H₇BrO₂
Storage Temperature: Ambient
CAS Number: 1191415-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR