2-Bromo-7-fluorobenzofuran ≥95%

Supplier: AOBChem USA

Warning

592324-5G 592324-500MG 592324-1G
77630-504EA 1822.07 USD
77630-504 77653-046 77643-842
2-Bromo-7-fluorobenzofuran ≥95%
2-Bromo-7-fluorobenzofuran

Precaución: For Research Use Only

Formula: C₈H₄BrFO
Storage Temperature: Ambient
CAS Number: 2149296-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR