1-Cyclohexyl-4-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

59211-5G 59211-25G 59211-1G 59211-500MG
77633-622EA 841 USD
77633-622 77656-652 77643-840 77651-790
1-Cyclohexyl-4-iodobenzene ≥95%
1-Cyclohexyl-4-iodobenzene

Precaución: For Research Use Only

Formula: C₁₂H₁₅I
Storage Temperature: Ambient
MDL Number: MFCD11111740
CAS Number: 91639-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR