Ethyl 3-(4-Bromophenyl)-2-cyanoacrylate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (E)-Ethyl 3-(4-bromophenyl)-2-cyanoacrylate

589849-5G 589849-25G 589849-1G 589849-500MG
77633-620EA 1155 USD
77633-620 77660-150 77643-836 77651-788
Ethyl 3-(4-Bromophenyl)-2-cyanoacrylate ≥95%
Ethyl 3-(4-Bromophenyl)-2-cyanoacrylate

Precaución: For Research Use Only

Formula: C₁₂H₁₀BrNO₂
MW: 280.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00176211
CAS Number: 18861-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR