Ethyl 3-(2-bromo-6-chlorophenyl)-3-hydroxypropanoate ≥97%

Supplier: AOBChem USA

Warning

58585-5G 58585-25G 58585-1G 58585-500MG
77635-224EA 594 USD
77635-224 77656-642 77643-830 77651-772
Ethyl 3-(2-bromo-6-chlorophenyl)-3-hydroxypropanoate ≥97%
Ethyl 3-(2-bromo-6-chlorophenyl)-3-hydroxypropanoate

Precaución: For Research Use Only

Formula: 307.568 g/mol
MW: C₁₁H₁₂BrClO₃
Storage Temperature: Ambient
MDL Number: MFCD34758716
CAS Number: 2755724-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR