2-((3-Fluorophenoxy)methyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

584804-5G 584804-1G 584804-25G 584804-500MG
77630-492EA 1484.23 USD
77630-492 77639-760 77660-142 77647-768
2-((3-Fluorophenoxy)methyl)benzonitrile ≥97%
2-((3-Fluorophenoxy)methyl)benzonitrile

Precaución: For Research Use Only

Formula: C₁₄H₁₀FNO
Storage Temperature: Ambient
CAS Number: 1019437-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR