(3-Bromo-5-(trifluoromethoxy)phenyl)(methyl)sulfane ≥95%
Supplier: AOBChem USA
|
Warning
|
58215-5G
58215-1G
58215-25G
58215-500MG
77635-220EA
390
USD
77635-220
77639-756
77656-636
77647-762
(3-Bromo-5-(trifluoromethoxy)phenyl)(methyl)sulfane ≥95%
(3-Bromo-5-(trifluoromethoxy)phenyl)(methyl)sulphane
Precaución: For Research Use Only
Formula:
C₈H₆BrF₃OS Storage Temperature: Ambient |
MDL Number:
MFCD32701833 CAS Number: 2167312-00-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...