1-(3-Chlorophenyl)-2,2-diethoxyethanone ≥95%

Supplier: AOBChem USA

Warning

580582-5G 580582-1G 580582-25G 580582-500MG
77630-486EA 1484.23 USD
77630-486 77639-754 77656-634 77647-760
1-(3-Chlorophenyl)-2,2-diethoxyethanone ≥95%
1-(3-Chlorophenyl)-2,2-diethoxyethanone

Precaución: For Research Use Only

Formula: C₁₂H₁₅ClO₃
Storage Temperature: Ambient
CAS Number: 1402458-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR