Ethyl 2-(2,6-difluoro-3-(methylthio)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

577209-5G 577209-1G 577209-500MG
77656-622EA 2281 USD
77656-622 77643-808 77651-742
Ethyl 2-(2,6-difluoro-3-(methylthio)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(2,6-difluoro-3-(methylthio)phenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: C₁₁H₁₀F₂O₃S
Storage Temperature: Ambient
CAS Number: 1898380-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR